(±)-1-Methyl-3-phenylpropylamine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Amino-1-phenylbutane, 3-Amino-1-phenyl-butane
OR5082-100G
OR5082-25G
OR5082-500G
APOSOR5082-100GEA
29.2
EUR
APOSOR5082-100G
APOSOR5082-25G
APOSOR5082-500G
(±)-1-Methyl-3-phenylpropylamine 98%
(±)-1-Methyl-3-phenylpropylamine
Formula:
C₁₀H₁₅N MW: 149,24 g/mol Boiling Pt: 220…222 °C Melting Pt: –50 °C Density: 0,928 g/cm³ (25 °C) Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008090 CAS Number: 22374-89-6 EINECS: 244-942-2 UN: 2922 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...