2,5-Diiodobenzonitrile ≥98%

Supplier: Apollo Scientific

OR510082-1G OR510082-250MG
APOSOR510082-1GEA 195 EUR
APOSOR510082-1G APOSOR510082-250MG
2,5-Diiodobenzonitrile ≥98%
2,5-Diiodobenzonitrile
Formula: C₇H₃I₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00463830
CAS Number: 79887-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR