Fluoranthene-3-boronic acid 98%

Supplier: Apollo Scientific

Synonyms: Fluoranthen-3-ylboronic acid

OR51016-5G
APOSOR51016-5GEA 186 EUR
APOSOR51016-5G
Fluoranthene-3-boronic acid 98%
Fluoranthene-3-boronic acid
Formula: C₁₆H₁₁BO₂
MW: 246,0683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660041
CAS Number: 359012-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR