Methyl propionylacetate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate
OR5113-500G
APOSOR5113-500GEA
88.6
EUR
APOSOR5113-500G
Methyl propionylacetate
Methyl propionylacetate
Formula:
C₆H₁₀O₃ MW: 130,14 g/mol Boiling Pt: 73…74 °C/5mmHg Melting Pt: –35 °C Density: 1,037 g/cm³ (25 °C) Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011705 CAS Number: 30414-53-0 EINECS: 250-184-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...