Fmoc-val-OSu 94%

Supplier: Apollo Scientific

Synonyms: N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu

OR51362-25G OR51362-100G
APOSOR51362-25GEA 37.7 EUR
APOSOR51362-25G APOSOR51362-100G
Fmoc-val-OSu 94%
Fmoc-val-OSu
Formula: C₂₄H₂₄N₂O₆
MW: 436.464 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153370
CAS Number: 130878-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR