1,2,3,4-Tetrahydroquinoline 98%
Supplier: Apollo Scientific
|
Danger
|
OR5166-100G
OR5166-25G
OR5166-500G
APOSOR5166-100GEA
35
EUR
APOSOR5166-100G
APOSOR5166-25G
APOSOR5166-500G
1,2,3,4-Tetrahydroquinoline 98%
1,2,3,4-Tetrahydroquinoline
Formula:
C₉H₁₁N MW: 133,19 g/mol Boiling Pt: 248…250 °C Melting Pt: 41883 °C Density: 1,059 g/cm³ (25 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006693 CAS Number: 635-46-1 EINECS: 211-237-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...