3-Chloro-5-hydroxybenzonitrile 98%

Supplier: Apollo Scientific
Warning

OR51727-5G OR51727-1G OR51727-500MG
APOSOR51727-5GEA 63.8 EUR
APOSOR51727-5G APOSOR51727-1G APOSOR51727-500MG
3-Chloro-5-hydroxybenzonitrile 98%
3-Chloro-5-hydroxybenzonitrile
Formula: C₇H₄ClNO
Melting Pt: 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11226540
CAS Number: 473923-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR