N,N-Dibutylformamide 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Formdibutylamide, Formic acid dibutylamide
OR51755-100G
OR51755-500G
APOSOR51755-100GEA
147
EUR
APOSOR51755-100G
APOSOR51755-500G
N,N-Dibutylformamide 99%
N,N-Dibutylformamide
Formula:
C₉H₁₉NO MW: 157,26 g/mol Boiling Pt: 120 °C/15mmHg Density: 0,864 g/cm³ (25 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003289 CAS Number: 761-65-9 EINECS: 212-090-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...