6-Chloro-2-pyridinecarboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde
OR51757-1G
OR51757-5G
APOSOR51757-1GEA
65.5
EUR
APOSOR51757-1G
APOSOR51757-5G
6-Chloro-2-pyridinecarboxaldehyde
6-Chloro-2-pyridinecarboxaldehyde
Formula:
C₆H₄ClNO MW: 141,55 g/mol Melting Pt: 68…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832941 CAS Number: 54087-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...