Methyl-5-hydroxypicolinate 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 5-hydroxypyridine-2-carboxylate
OR51766-1G
OR51766-5G
APOSOR51766-1GEA
29
EUR
APOSOR51766-1G
APOSOR51766-5G
Methyl-5-hydroxypicolinate 95+%
Methyl 5-hydroxypicolinate
Formula:
C₇H₇NO₃ MW: 153,1354 g/mol Melting Pt: 191…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD05256426 CAS Number: 30766-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...