Methyl-3-hydroxyisonicotinate 98%

Supplier: Apollo Scientific
Warning

OR51771-1G OR51771-10G OR51771-5G
APOSOR51771-1GEA 98.6 EUR
APOSOR51771-1G APOSOR51771-10G APOSOR51771-5G
Methyl-3-hydroxyisonicotinate 98%
Methyl 3-hydroxyisonicotinate
Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00661299
CAS Number: 10128-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR