3-Cyclopentylacrylonitrile 97%

Supplier: Apollo Scientific
Danger

OR51789-100G OR51789-25G
APOSOR51789-100GEA 266 EUR
APOSOR51789-100G APOSOR51789-25G
3-Cyclopentylacrylonitrile 97%
3-Cyclopentylacrylonitrile
Formula: C₈H₁₁N
MW: 121.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11857755
CAS Number: 591769-05-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR