3'-Iodoacetophenone

Supplier: Apollo Scientific
Danger

Synonyms: 1-(3-Iodophenyl)ethanone, m-Iodoacetophenone, 1-(3-Iodophenyl)ethan-1-one

OR51806-100G OR51806-25G
APOSOR51806-100GEA 389 EUR
APOSOR51806-100G APOSOR51806-25G
3'-Iodoacetophenone
3'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246,05 g/mol
Boiling Pt: 107 °C/0,75mmHg
Density: 1,767 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060283
CAS Number: 14452-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR