2-Bromo-5-chloropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
OR5182-100G
OR5182-10G
OR5182-25G
APOSOR5182-100GEA
59.7
EUR
APOSOR5182-100G
APOSOR5182-10G
APOSOR5182-25G
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Formula:
C₅H₃BrClN Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234006 CAS Number: 40473-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...