4-(Chloromethyl)styrene, Technical Grade
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride
OR51822-250G
OR51822-25G
APOSOR51822-250GEA
364
EUR
APOSOR51822-250G
APOSOR51822-25G
4-(Chloromethyl)styrene, Technical Grade
4-(Chloromethyl)styrene
Formula:
C₉H₉Cl MW: 152,62 g/mol Boiling Pt: 229 °C Density: 1,083 g/cm³ (25 °C) Flash Pt: 104 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051362 CAS Number: 1592-20-7 EINECS: 216-471-2 UN: 2922 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...