Methyl-2-bromonicotinate

Supplier: Apollo Scientific

OR51917-1G OR51917-25G OR51917-5G
APOSOR51917-1GEA 21.9 EUR
APOSOR51917-1G APOSOR51917-25G APOSOR51917-5G
Methyl-2-bromonicotinate
Methyl 2-bromonicotinate
Formula: C₇H₆BrNO₂
Melting Pt: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD07367915
CAS Number: 52718-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR