4-Bromobenzophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Bromophenyl)(phenyl)methanone, p-Bromobenzophenone

OR51939-500G
APOSOR51939-500GEA 261 EUR
APOSOR51939-500G
4-Bromobenzophenone 98%
4-Bromobenzophenone
Formula: C₁₃H₉BrO
MW: 261,12 g/mol
Boiling Pt: 350 °C
Melting Pt: 79…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000103
CAS Number: 90-90-4
EINECS: 202-024-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR