N-Boc-Piperidin-4-ylacetic acid methyl ester

Supplier: Apollo Scientific
Warning

Synonyms: N-Boc-Piperidin-4-yl-acetic acid methyl ester, Methyl (piperidin-4-yl)acetate, N-BOC protected

OR51981-25G OR51981-5G
APOSOR51981-25GEA 144 EUR
APOSOR51981-25G APOSOR51981-5G
N-Boc-Piperidin-4-ylacetic acid methyl ester
N-Boc-Piperidin-4-ylacetic acid methyl ester
Formula: C₁₃H₂₃NO₄
MW: 257,3300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632529
CAS Number: 175213-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR