6-Phenyl-3-formylpyridine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Phenyl-5-pyridinecarboxaldehyde, 6-Phenylnicotinaldehyde

OR5202-1G OR5202-250MG
APOSOR5202-1GEA 329 EUR
APOSOR5202-1G APOSOR5202-250MG
6-Phenyl-3-formylpyridine 97%
6-Phenyl-3-formylpyridine
Formula: C₁₂H₉NO
MW: 183.21 g/mol
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD03086130
CAS Number: 63056-20-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR