2-Amino-3-iodophenol 98%
Supplier: Apollo Scientific
|
Warning
|
OR52073-1G
OR52073-5G
APOSOR52073-1GEA
510
EUR
APOSOR52073-1G
APOSOR52073-5G
2-Amino-3-iodophenol 98%
2-Amino-3-iodophenol
Formula:
C₆H₆INO Melting Pt: 137 °C Storage Temperature: Ambient |
CAS Number:
443921-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...