4-Bromo-2-iodophenol 98%

Supplier: Apollo Scientific
Warning

OR52087-1G OR52087-5G
APOSOR52087-1GEA 40.8 EUR
APOSOR52087-1G APOSOR52087-5G
4-Bromo-2-iodophenol 98%
4-Bromo-2-iodophenol
Formula: C₆H₄BrIO
MW: 298,91 g/mol
Melting Pt: 71,8…72.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09029644
CAS Number: 207115-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR