3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid

OR52107-1G OR52107-250MG OR52107-5G
APOSOR52107-1GEA 136 EUR
APOSOR52107-1G APOSOR52107-250MG APOSOR52107-5G
3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid
3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid
Formula: C₈H₅BrN₂O₂
MW: 241,05 g/mol
Melting Pt: 282…285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD21602377
CAS Number: 1315360-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR