6-Methoxyisatin
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Methoxyindoline-2,3-dione
OR52220-10G
OR52220-1G
OR52220-5G
APOSOR52220-10GEA
179
EUR
APOSOR52220-10G
APOSOR52220-1G
APOSOR52220-5G
6-Methoxyisatin
6-Methoxyisatin
Formula:
C₉H₇NO₃ MW: 177,1568 g/mol Melting Pt: 223…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00667731 CAS Number: 52351-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...