6-Chloro-1-tetralone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-3,4-dihydronaphthalen-1(2H)-one, 6-Chloro-3,4-dihydro-1(2H)-naphthalenone

OR52290-1G OR52290-5G OR52290-10G
APOSOR52290-1GEA 21.9 EUR
APOSOR52290-1G APOSOR52290-5G APOSOR52290-10G
6-Chloro-1-tetralone 98%
6-Chloro-1-tetralone
Formula: C₁₀H₉ClO
MW: 180,63 g/mol
Boiling Pt: 150 °C/0,75mmHg
Melting Pt: 31…32 °C
Density: 1,24 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04114377
CAS Number: 26673-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR