2-Chlorocyclopentanone 98%
Supplier: Apollo Scientific
OR5230-1G
OR5230-5G
APOSOR5230-1GEA
66.7
EUR
APOSOR5230-1G
APOSOR5230-5G
2-Chlorocyclopentanone 98%
2-Chlorocyclopentanone
Formula:
C₅H₇ClO MW: 118.56 g/mol Boiling Pt: 72…74 °C/12mmHg Density: 1.185 g/cm³ (25 °C) Flash Pt: 77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001410 CAS Number: 694-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...