2-tert-Butyl-1H-imidazole

Supplier: Apollo Scientific
Warning

Synonyms: 2-(tert-Butyl)-1H-imidazole

OR52331-5G
APOSOR52331-5GEA 77 EUR
APOSOR52331-5G
2-tert-Butyl-1H-imidazole
2-tert-Butyl-1H-imidazole
Formula: C₇H₁₂N₂
MW: 124,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618480
CAS Number: 36947-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR