6-(Methoxycarbonyl)picolinic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,6-Pyridinedicarboxylic acid monomethyl ester, Dipicolinic acid monomethyl ester, Monomethyl 2,6-Pyridinedicarboxylate, Monomethyl dipicolinate
OR52341-10G
OR52341-5G
APOSOR52341-10GEA
88.7
EUR
APOSOR52341-10G
APOSOR52341-5G
6-(Methoxycarbonyl)picolinic acid 97%
6-(Methoxycarbonyl)picolinic acid
Formula:
C8H7NO4 MW: 181,1455 g/mol Melting Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00806109 CAS Number: 7170-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...