(1S,2S)-(+)-1-Amino-2-indanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(1S,2S)-1-Amino-2-indanol, (1S,2S)-(+)-1-Amino-2-hydroxyindan
OR52536-5G
APOSOR52536-5GEA
159
EUR
APOSOR52536-5G
(1S,2S)-(+)-1-Amino-2-indanol
(1S,2S)-(+)-1-Amino-2-indanol
Formula:
C₉H₁₁NO MW: 149,19 g/mol Melting Pt: 142 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275383 CAS Number: 163061-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...