4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO)
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy
OR52537-25G
APOSOR52537-25GEA
142
EUR
APOSOR52537-25G
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO)
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-Acetamido-TEMPO, free radical)
Formula:
C₁₁H₂₁N₂O₂ MW: 213,3 g/mol Melting Pt: 143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043593 CAS Number: 14691-89-5 EINECS: 423-840-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...