trans-N-Fmoc-4-tert-butoxy-L-proline 95%

Supplier: Apollo Scientific
Warning

Synonyms: N-Fmoc-4-trans-(tert-butoxy)-L-proline, Fmoc-Hyp(tBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline, (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidine-2-carboxylic acid

OR52599-1G OR52599-5G
APOSOR52599-1GEA 21.9 EUR
APOSOR52599-1G APOSOR52599-5G
trans-N-Fmoc-4-tert-butoxy-L-proline 95%
trans-N-Fmoc-4-tert-butoxy-L-proline
Formula: C₂₄H₂₇NO₅
MW: 409,482 g/mol
Melting Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151930
CAS Number: 122996-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR