9-(4-Bromophenyl)carbazole

Supplier: Apollo Scientific
Warning

OR52631-25G OR52631-5G
APOSOR52631-25GEA 71.3 EUR
APOSOR52631-25G APOSOR52631-5G
9-(4-Bromophenyl)carbazole
9-(4-Bromophenyl)carbazole
Formula: C₁₈H₁₂BrN
MW: 322,21 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD11617969
CAS Number: 57102-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR