Etilefrine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride
OR52664-25G
OR52664-5G
APOSOR52664-25GEA
619
EUR
APOSOR52664-25G
APOSOR52664-5G
Etilefrine hydrochloride
Etilefrine hydrochloride
Formula:
C₁₀H₁₅NO₂ MW: 217,69 g/mol Melting Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060167 CAS Number: 943-17-9 EINECS: 213-398-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...