Trimesic acid 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
OR52672-100G
OR52672-500G
APOSOR52672-100GEA
68.3
EUR
APOSOR52672-100G
APOSOR52672-500G
Trimesic acid 95%
Trimesic acid
Formula:
C₉H₆O₆ MW: 210,14 g/mol Melting Pt: 374…376 °C Flash Pt: 328 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002517 CAS Number: 554-95-0 EINECS: 209-077-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...