H-Deg-OH

Supplier: Apollo Scientific
Warning

OR52678-1G OR52678-5G
APOSOR52678-1GEA 82.8 EUR
APOSOR52678-1G APOSOR52678-5G
H-Deg-OH
H-Deg-OH
Formula: C₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01737173
CAS Number: 2566-29-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR