Methoxyacetonitrile
Supplier: Apollo Scientific
|
Warning
|
OR5269-100G
OR5269-25G
OR5269-5G
APOSOR5269-100GEA
786
EUR
APOSOR5269-100G
APOSOR5269-25G
APOSOR5269-5G
Methoxyacetonitrile
Methoxyacetonitrile
Formula:
C₃H₅NO MW: 71,08 g/mol Boiling Pt: 118…119 °C/731mmHg Density: 0,956 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001892 CAS Number: 1738-36-9 EINECS: 217-092-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...