1-(2-Bromophenyl)piperidine 96%

Supplier: Apollo Scientific

OR52750-25G OR52750-5G
APOSOR52750-25GEA 324 EUR
APOSOR52750-25G APOSOR52750-5G
1-(2-Bromophenyl)piperidine 96%
1-(2-Bromophenyl)piperidine
Formula: C₁₁H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD04112586
CAS Number: 156808-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR