1-(3-Bromopyridin-2-yl)ethanone 97%

Supplier: Apollo Scientific

Synonyms: 2-Acetyl-3-bromopyridine

OR52783-5G
APOSOR52783-5GEA 87 EUR
APOSOR52783-5G
1-(3-Bromopyridin-2-yl)ethanone 97%
1-(3-Bromopyridin-2-yl)ethanone
Formula: C₇H₆BrNO
MW: 200,0326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257141
CAS Number: 111043-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR