4-Amino-2-chlorobenzonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-4-cyanoaniline

OR5293-25G OR5293-5G
APOSOR5293-25GEA 46.4 EUR
APOSOR5293-25G APOSOR5293-5G
4-Amino-2-chlorobenzonitrile 98%
4-Amino-2-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3
EINECS: 244-114-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR