3-Amino-3-oxopropanoic acid
Supplier: Apollo Scientific
Synonyms:
MALONAMIC acid
OR52961-250MG
APOSOR52961-250MGEA
195
EUR
APOSOR52961-250MG
3-Amino-3-oxopropanoic acid
3-Amino-3-oxopropanoic acid
Formula:
C₃H₅NO₃ MW: 103,08 g/mol Melting Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00154605 CAS Number: 2345-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...