Dichloroacetyl chloride 97%

Supplier: Apollo Scientific
Danger

OR5297-100G OR5297-500G
APOSOR5297-100GEA 24.8 EUR
APOSOR5297-100G APOSOR5297-500G
Dichloroacetyl chloride 97%
Dichloroacetyl chloride
Formula: C₂HCl₃O
Boiling Pt: 107…108 °C
Density: 1,532 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000840
CAS Number: 79-36-7
EINECS: 201-199-9
UN: 1765
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR