1-Iodopentane 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Pentyl iodide

OR53033-100G OR53033-500G
APOSOR53033-100GEA 120 EUR
APOSOR53033-100G APOSOR53033-500G
1-Iodopentane 98%
1-Iodopentane
Formula: C₅H₁₁I
Boiling Pt: 154…156 °C
Melting Pt: –86 °C
Density: 1,515 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001100
CAS Number: 628-17-1
EINECS: 211-030-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR