Propyl acetate 99%

Supplier: Apollo Scientific

OR5312-1G OR5312-5G
APOSOR5312-1GEA 68.4 EUR
APOSOR5312-1G APOSOR5312-5G
Propyl acetate 99%
Propyl acetate
Formula: C₅H₁₀O₂
Boiling Pt: 99…102 °C
Melting Pt: –92 °C
Density: 0,887 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009372
CAS Number: 109-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR