(4-Bromo-2-pyridinyl)methanol 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-(hydroxymethyl)pyridine, 4-Bromo-2-pyridinemethanol, (4-Bromopyridin-2-yl)methanol

OR53132-1G OR53132-5G
APOSOR53132-1GEA 0 EUR
APOSOR53132-1G APOSOR53132-5G
(4-Bromo-2-pyridinyl)methanol 95%
(4-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
MW: 188,0200 g/mol
Melting Pt: 49…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09800557
CAS Number: 131747-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR