Fmoc-O-trityl-L-threonine 98.5+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Fmoc-Thr(Trt)-OH, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(trityloxy)butanoic acid
OR53138-1G
OR53138-25G
OR53138-5G
APOSOR53138-1GEA
132
EUR
APOSOR53138-1G
APOSOR53138-25G
APOSOR53138-5G
Fmoc-O-trityl-L-threonine 98.5+%
Fmoc-O-trityl-L-threonine
Formula:
C₃₈H₃₃NO₅ MW: 583,684 g/mol Melting Pt: 70…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00237034 CAS Number: 133180-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...