6-Chloro-1-hexene

Supplier: Apollo Scientific
Warning

OR53170-25G OR53170-5G
APOSOR53170-25GEA 270 EUR
APOSOR53170-25G APOSOR53170-5G
6-Chloro-1-hexene
6-Chloro-1-hexene
Formula: C₆H₁₁Cl
MW: 118,61 g/mol
Boiling Pt: 135…136 °C
Density: 0,896 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039391
CAS Number: 928-89-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR