2-Iodobutane 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Butyl iodide, sec-Butyl iodide
OR5340-100G
OR5340-500G
APOSOR5340-100GEA
58
EUR
APOSOR5340-100G
APOSOR5340-500G
2-Iodobutane 98%
2-Iodobutane
Formula:
C₄H₉I MW: 184,02 g/mol Boiling Pt: 119…120 °C Melting Pt: –104 °C Density: 1,598 g/cm³ (25 °C) Flash Pt: 21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001072 CAS Number: 513-48-4 EINECS: 208-163-1 UN: 2390 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...