4-(Methoxycarbonyl)phenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid
OR5354-100G
OR5354-25G
OR5354-5G
APOSOR5354-100GEA
108
EUR
APOSOR5354-100G
APOSOR5354-25G
APOSOR5354-5G
4-(Methoxycarbonyl)phenylboronic acid 98%
4-(Methoxycarbonyl)phenylboronic acid
Formula:
C₈H₉BO₄ MW: 179.97 g/mol Melting Pt: 197…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632203 CAS Number: 99768-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...