1,2,4-Trimethylbenzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: Pseudocumene

OR5391-500G OR5391-2KG OR5391-5KG
APOSOR5391-500GEA 33.5 EUR
APOSOR5391-500G APOSOR5391-2KG APOSOR5391-5KG
1,2,4-Trimethylbenzene 98%
1,2,4-Trimethylbenzene
Formula: C₉H₁₂
Boiling Pt: 168 °C
Melting Pt: –44 °C
Density: 0,876 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR