4-Iodophenol 97%

Supplier: Apollo Scientific
Danger

OR5400-25G OR5400-100G OR5400-10G
APOSOR5400-25GEA 61.1 EUR
APOSOR5400-25G APOSOR5400-100G APOSOR5400-10G
4-Iodophenol 97%
4-Iodophenol
Formula: C₆H₅IO
MW: 220,01 g/mol
Boiling Pt: 138 °C/5mmHg
Melting Pt: 91…95 °C
Density: 1,857 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR