3-Iodo-p-anisidine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Iodo-4-methoxyaniline
OR5428-1G
OR5428-25G
OR5428-5G
APOSOR5428-1GEA
23.2
EUR
APOSOR5428-1G
APOSOR5428-25G
APOSOR5428-5G
3-Iodo-p-anisidine
3-Iodo-p-anisidine
Formula:
C₇H₈INO MW: 249,0490 g/mol Melting Pt: 74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06656566 CAS Number: 74587-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...